missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Hydroxy-3-nitropyridine, 95%

Brand:  Alfa Aesar™ B20963.22

Additional Details : CAS : 5435-54-1 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11490027

  • 369.00€ / 100g


In stock, ready to ship: 3
Add to basket

Description

Description

Specifications

Chemical Identifiers

5435-54-1
140.098
YUWOLBZMQDGRFV-UHFFFAOYSA-N
228506
C1=CNC=C(C1=O)[N+](=O)[O-]
C5H4N2O3
MFCD00128874
4-hydroxy-3-nitropyridine, 3-nitropyridin-4-ol, 3-nitro-4-pyridinol, 3-nitro-4-hydroxypyridine, 3-nitropyridin-4 1h-one, 3-nitro-4-pyridone, 4-pyridinol, 3-nitro, 4 1h-pyridinone, 3-nitro, 3-nitro-1,4-dihydropyridin-4-one, 4-hydroxy-3-nitropytidine
3-nitro-1H-pyridin-4-one

Specifications

4-Hydroxy-3-nitropyridine
100g
C5H4N2O3
1072928
YUWOLBZMQDGRFV-UHFFFAOYSA-N
3-nitro-1H-pyridin-4-one
228506
95%
~280°C (decomposition)
5435-54-1
MFCD00128874
4-hydroxy-3-nitropyridine, 3-nitropyridin-4-ol, 3-nitro-4-pyridinol, 3-nitro-4-hydroxypyridine, 3-nitropyridin-4 1h-one, 3-nitro-4-pyridone, 4-pyridinol, 3-nitro, 4 1h-pyridinone, 3-nitro, 3-nitro-1,4-dihydropyridin-4-one, 4-hydroxy-3-nitropytidine
C1=CNC=C(C1=O)[N+](=O)[O-]
140.098
140.1
Videos
SDS Documents