missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-7021299
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
123-08-0
C7H6O2
122.123
MFCD00006939
RGHHSNMVTDWUBI-UHFFFAOYSA-N
p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde
126
CHEBI:17597
4-hydroxybenzaldehyde
C1=CC(=CC=C1C=O)O
This item is not returnable. View return policy
123-08-0
C7H6O2
122.123
MFCD00006939
RGHHSNMVTDWUBI-UHFFFAOYSA-N
p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde
126
CHEBI:17597
4-hydroxybenzaldehyde
C1=CC(=CC=C1C=O)O

4-Hydroxybenzaldehyde is used as a chemical that maintains bactericidal activity when tested against certain bacteria strains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzaldehyde is used as a chemical that maintains bactericidal activity when tested against certain bacteria strains.

Solubility
Soluble in water(13.8g/L), ether, alcohol, and acetone.

Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Melting Point 115°C to 119°C
Density 1.129
Boiling Point 191°C (50 mmHg)
Flash Point 174°C (345°F)
Quantity 25 g
Beilstein 471352
Sensitivity Air Sensitive
Merck Index 14,4811
Solubility Information Soluble in water(13.8g/L),ether,alcohol,and acetone.
Formula Weight 122.12
Percent Purity 98%
Chemical Name or Material 4-Hydroxybenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'