missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxyflavanone, TRC

Product Code. 30390858
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390858 250 mg 250mg
30395424 50 mg -
30405837 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390858 Brand Toronto Research Chemicals Brand No. TRCH942443250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6515-37-3
Molecular Formula C15 H12 O3
Molecular Weight (g/mol) 240.25
Synonym (±)-4'-Hydroxyflavanone
IUPAC Name 2-(4-hydroxyphenyl)chroman-4-one
SMILES Oc1ccc(cc1)C2CC(=O)c3ccccc3O2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H12O3/c16-11-7-5-10(6-8-11)15-9-13(17)12-3-1-2-4-14(12)18-15/h1-8,15-16H,9H2
Formula Weight 240.0786
Chemical Name or Material 4'-Hydroxyflavanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.