missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)benzonitrile, 97%, Thermo Scientific Chemicals

Catalog Number p-7077498
36.80 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
874-89-5
C8H7NO
133.15
MFCD00870633
XAASLEJRGFPHEV-UHFFFAOYSA-N
4-hydroxymethyl benzonitrile, 4-cyanobenzyl alcohol, p-cyanobenzyl alcohol, ccris 5116, benzonitrile, 4-hydroxymethyl, 4-cyanobenzenemethanol, 4-cyanophenyl methanol, 4-hydroxymethyl-benzonitrile, 4 hydroxymethyl-benzonitrile
160549
4-(hydroxymethyl)benzonitrile
OCC1=CC=C(C=C1)C#N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
874-89-5
C8H7NO
133.15
MFCD00870633
XAASLEJRGFPHEV-UHFFFAOYSA-N
4-hydroxymethyl benzonitrile, 4-cyanobenzyl alcohol, p-cyanobenzyl alcohol, ccris 5116, benzonitrile, 4-hydroxymethyl, 4-cyanobenzenemethanol, 4-cyanophenyl methanol, 4-hydroxymethyl-benzonitrile, 4 hydroxymethyl-benzonitrile
160549
4-(hydroxymethyl)benzonitrile
OCC1=CC=C(C=C1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 41.0°C to 42.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 133.15
Percent Purity 97%
Chemical Name or Material 4-(Hydroxymethyl)benzonitrile
compliance-icons
Product Identifier
  • 4-(Hydroxymethyl)benzonitrile, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'