missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzenesulfonamide, 95%, Thermo Scientific Chemicals

Catalog Number p-7077574
27.26 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
825-86-5
C6H6INO2S
283.09
KQZFABSTXSNEQH-UHFFFAOYSA-N
p-iodobenzenesulfonamide, 4-iodobenzenesulphonamide, 4-iodobenzene-1-sulfonamide, benzenesulfonamide,4-iodo, benzenesulfonamide, p-iodo, p-jodbenzolsulfonamid, 4-iodibenzenesulphonamide, 4-iodo-benzenesulfonamide, 4-iodo-1-sulfamoylbenzene, 3-11-00-00110 beilstein handbook reference
13218
4-iodobenzenesulfonamide
C1=CC(=CC=C1S(=O)(=O)N)I
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
825-86-5
C6H6INO2S
283.09
KQZFABSTXSNEQH-UHFFFAOYSA-N
p-iodobenzenesulfonamide, 4-iodobenzenesulphonamide, 4-iodobenzene-1-sulfonamide, benzenesulfonamide,4-iodo, benzenesulfonamide, p-iodo, p-jodbenzolsulfonamid, 4-iodibenzenesulphonamide, 4-iodo-benzenesulfonamide, 4-iodo-1-sulfamoylbenzene, 3-11-00-00110 beilstein handbook reference
13218
4-iodobenzenesulfonamide
C1=CC(=CC=C1S(=O)(=O)N)I

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 193.0°C to 195.0°C
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Linear Formula IC6H4SO2NH2
Quantity 1 g
Formula Weight 283.09
Percent Purity 95%
Chemical Name or Material 4-Iodobenzenesulfonamide
compliance-icons
Product Identifier
  • 4-Iodobenzenesulfonamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'