Learn More
Alfa Aesar™ 4-Isopropoxybenzeneboronic acid, 97%
Brand: Alfa Aesar™ H52675.06
Additional Details : CAS : 153624-46-5 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
153624-46-5 | |
180.01 | |
CJUHQADBFQRIMC-UHFFFAOYSA-N | |
3698726 | |
B(C1=CC=C(C=C1)OC(C)C)(O)O |
C9H13BO3 | |
MFCD03427051 | |
4-isopropoxyphenylboronic acid, 4-isopropoxylphenylboronic acid, 4-isopropoxyphenyl boronic acid, 4-isopropoxybenzeneboronic acid, 4-isopropyloxyphenylboronic acid, 4-propan-2-yloxyphenyl boronic acid, boronic acid, 4-1-methylethoxy phenyl, 4-propan-2-yloxy phenyl boronic acid, 4-iso-propoxyphenylboronic acid, p-isopropoxyphenylboronic acid | |
(4-propan-2-yloxyphenyl)boronic acid |
Specifications
4-Isopropoxybenzeneboronic acid | |
5g | |
C9H13BO3 | |
4-isopropoxyphenylboronic acid, 4-isopropoxylphenylboronic acid, 4-isopropoxyphenyl boronic acid, 4-isopropoxybenzeneboronic acid, 4-isopropyloxyphenylboronic acid, 4-propan-2-yloxyphenyl boronic acid, boronic acid, 4-1-methylethoxy phenyl, 4-propan-2-yloxy phenyl boronic acid, 4-iso-propoxyphenylboronic acid, p-isopropoxyphenylboronic acid | |
B(C1=CC=C(C=C1)OC(C)C)(O)O | |
180.01 | |
180.01 |
150°C to 154°C | |
153624-46-5 | |
MFCD03427051 | |
CJUHQADBFQRIMC-UHFFFAOYSA-N | |
(4-propan-2-yloxyphenyl)boronic acid | |
3698726 | |
97% |