missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trihydroxybenzene, 97%

Product Code. 11414384
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11414384

Brand: Thermo Scientific Alfa Aesar A13069.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 533-73-3 | C6H6O3 | 126.111 g/mol

1,2,4-Trihydroxybenzene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.It is used as a metabolite of benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4-Trihydroxybenzene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.It is used as a metabolite of benzene.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acid chlorides, acid anhydrides, bases, oxidizing agents. Air & Light Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-73-3
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00002198
InChI Key GGNQRNBDZQJCCN-UHFFFAOYSA-N
Synonym 1,2,4-benzenetriol, 1,2,4-trihydroxybenzene, hydroxyhydroquinone, hydroxyquinol, oxyhydroquinone, 2,5-dihydroxyphenol, oxyhydrochinon, hydroquinone, hydroxy, 4-hydroxycatechol, 1,3,4-trihydroxybenzene
PubChem CID 10787
ChEBI CHEBI:16971
IUPAC Name benzene-1,2,4-triol
SMILES C1=CC(=C(C=C1O)O)O

Specifications

Melting Point ∼140°C (sublimation)
Quantity 25 g
Beilstein 2042863
Sensitivity Air and light sensitive
Merck Index 14,1073
Solubility Information Soluble in water.
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material 1,2,4-Trihydroxybenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.