missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2(5H)-furanone, 97%, Thermo Scientific Chemicals

Catalog Number p-3758302
29.86 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
69556-70-3
C5H6O3
114.1
MFCD00071565
VOYDEHILKLSVNN-UHFFFAOYSA-N
643477
3-methoxy-2H-furan-5-one
COC1=CC(=O)OC1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
69556-70-3
C5H6O3
114.1
MFCD00071565
VOYDEHILKLSVNN-UHFFFAOYSA-N
643477
3-methoxy-2H-furan-5-one
COC1=CC(=O)OC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 60°C to 65°C
Color Yellow
Boiling Point 130°C to 131°C (18.0 mbar)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Synonym 4-methoxy-2 5h-furanone, 4-methoxyfuran-2 5h-one, 4-methoxy-5h-furan-2-one, 2 5h-furanone,4-methoxy, 2 5h-furanone, 4-methoxy, 4-methoxy-5-hydrofuran-2-one
Solubility Information Solubility in water: 240g/L (20°C)
Formula Weight 114.1
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 4-Methoxy-2(5H)-furanone
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxy-2(5H)-furanone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'