missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-2-methyl-N-(2-pyridyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
4-Methoxy-2-methyl-N-(2-pyridyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
Specifications
Specifications
| Chemical Name or Material | 4-Methoxy-2-methyl-N-(2-pyridyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide |
| CAS | 76780-03-5 |
| Analyte or Component Names | 4-Methoxy-2-methyl-N-(2-pyridyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H15 N3 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-Methoxy-2-methyl-N-(2-pyridyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide |
| InChI Formula | InChI=1S/C16H15N3O4S/c1-19-14(16(20)18-13-9-5-6-10-17-13)15(23-2)11-7-3-4-8-12(11)24(19,21)22/h3-10H,1-2H3,(H,17,18,20) |
| SMILES | COC1=C(N(C)S(=O)(=O)c2ccccc12)C(=O)Nc3ccccn3 |
| IUPAC Name | 4-methoxy-2-methyl-1,1-dioxo-N-pyridin-2-yl-1λ^{6},2-benzothiazine-3-carboxamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction