Learn More
Alfa Aesar™ 4-Methoxy-2-(trifluoromethyl)benzaldehyde, 98%
Brand: Alfa Aesar™ H26123.MD
Additional Details : CAS : 106312-36-1 Weight : 0.00025kg
Quantity | 250mg |
---|
Chemical Identifiers
106312-36-1 | |
204.148 | |
BVPVUMRIGHMFNV-UHFFFAOYSA-N | |
2778286 | |
COC1=CC(=C(C=C1)C=O)C(F)(F)F |
C9H7F3O2 | |
MFCD01091011 | |
4-methoxy-2-trifluoromethyl benzaldehyde, benzaldehyde, 4-methoxy-2-trifluoromethyl, 4-formyl-3-trifluoromethyl anisole, pubchem1463, acmc-1c5gx, ksc504i7t, 2-formyl-5-methoxybenzotrifluoride, 4-methoxy-2-trifluoromethyl-benzaldehyde, benzaldehyde,4-methoxy-2-trifluoromethyl | |
4-methoxy-2-(trifluoromethyl)benzaldehyde |
Specifications
4-Methoxy-2-(trifluoromethyl)benzaldehyde | |
39°C to 41°C | |
106312-36-1 | |
MFCD01091011 | |
BVPVUMRIGHMFNV-UHFFFAOYSA-N | |
4-methoxy-2-(trifluoromethyl)benzaldehyde | |
2778286 | |
98% |
Air Sensitive | |
250mg | |
C9H7F3O2 | |
4-methoxy-2-trifluoromethyl benzaldehyde, benzaldehyde, 4-methoxy-2-trifluoromethyl, 4-formyl-3-trifluoromethyl anisole, pubchem1463, acmc-1c5gx, ksc504i7t, 2-formyl-5-methoxybenzotrifluoride, 4-methoxy-2-trifluoromethyl-benzaldehyde, benzaldehyde,4-methoxy-2-trifluoromethyl | |
COC1=CC(=C(C=C1)C=O)C(F)(F)F | |
204.148 | |
204.15 |