missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-buten-2-one, tech. 90%, Thermo Scientific Chemicals

Catalog Number p-7081308
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
4652-27-1
C5H8O2
100.12
MFCD00008778,MFCD00008778,MFCD00008778
VLLHEPHWWIDUSS-ARJAWSKDSA-N
z-4-methoxybut-3-en-2-one, 3z-4-methoxybut-3-en-2-one, 3-buten-2-one, 4-methoxy-, 3z, z-4-methoxy-3-butene-2-one
643837
(Z)-4-methoxybut-3-en-2-one
CO\C=C/C(C)=O
This item is not returnable. View return policy
4652-27-1
C5H8O2
100.12
MFCD00008778,MFCD00008778,MFCD00008778
VLLHEPHWWIDUSS-ARJAWSKDSA-N
z-4-methoxybut-3-en-2-one, 3z-4-methoxybut-3-en-2-one, 3-buten-2-one, 4-methoxy-, 3z, z-4-methoxy-3-butene-2-one
643837
(Z)-4-methoxybut-3-en-2-one
CO\C=C/C(C)=O

4-Methoxy-3-buten-2-one is an important precursor for the synthesis of 1-methoxy-3-trimethylsilyloxy-1,3-butadiene and the Danishefsky diene and its equivalent. It is widely used as a reactant in the synthesis of hydroisoquinoline derivatives. It is employed as an intermediate for the synthesis of manzamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-3-buten-2-one is an important precursor for the synthesis of 1-methoxy-3-trimethylsilyloxy-1,3-butadiene and the Danishefsky diene and its equivalent. It is widely used as a reactant in the synthesis of hydroisoquinoline derivatives. It is employed as an intermediate for the synthesis of manzamine.

Solubility
Miscible with tetrahydrofuran, ether, acetonitrile and benzene.

Notes
Keep away from sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Density 0.99
Boiling Point 66°C to 69°C (12 mmHg)
Flash Point 63°C (145°F)
Refractive Index 1.465
Quantity 10 g
Beilstein 1741053
Solubility Information Miscible with tetrahydrofuran,ether,acetonitrile and benzene.
Formula Weight 100.12
Percent Purity 90%
Grade Technical
Chemical Name or Material 4-Methoxy-3-buten-2-one
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'