missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methoxy-3-methyl-5-(trifluoromethyl)aniline, 97%

Brand:  Alfa Aesar™ H35270.06

Additional Details : Weight : 0.00500kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15407497

  • 508.00€ / 5g


Estimated Shipment
26-04-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

1431329-84-8
205.18
ZFHSTWVPCFNUGW-UHFFFAOYSA-N
86277688
CC1=CC(=CC(=C1OC)C(F)(F)F)N
C9H10F3NO
MFCD22682815
4-methoxy-3-methyl-5-trifluoromethyl aniline
4-methoxy-3-methyl-5-(trifluoromethyl)aniline

Specifications

4-Methoxy-3-methyl-5-(trifluoromethyl)aniline
1431329-84-8
MFCD22682815
4-methoxy-3-methyl-5-trifluoromethyl aniline
CC1=CC(=CC(=C1OC)C(F)(F)F)N
205.18
205.18
5g
C9H10F3NO
UN2810
ZFHSTWVPCFNUGW-UHFFFAOYSA-N
4-methoxy-3-methyl-5-(trifluoromethyl)aniline
86277688
97%
Videos
SDS Documents