missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-methyl-5-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Catalog Number p-7032887
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1431329-84-8
C9H10F3NO
205.18
MFCD22682815
ZFHSTWVPCFNUGW-UHFFFAOYSA-N
4-methoxy-3-methyl-5-trifluoromethyl aniline
86277688
4-methoxy-3-methyl-5-(trifluoromethyl)aniline
COC1=C(C=C(N)C=C1C)C(F)(F)F
This item is not returnable. View return policy
1431329-84-8
C9H10F3NO
205.18
MFCD22682815
ZFHSTWVPCFNUGW-UHFFFAOYSA-N
4-methoxy-3-methyl-5-trifluoromethyl aniline
86277688
4-methoxy-3-methyl-5-(trifluoromethyl)aniline
COC1=C(C=C(N)C=C1C)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN2810
Formula Weight 205.18
Percent Purity 97%
Chemical Name or Material 4-Methoxy-3-methyl-5-(trifluoromethyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'