missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyindole-2-carboxylic acid, 97+%, Thermo Scientific Chemicals

Catalog Number 15409097
372.01 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
103260-65-7
C10H9NO3
191.186
MFCD02664458
ZZAVIQXQBBOHBB-UHFFFAOYSA-N
4-methoxyindole-2-carboxylic acid, 4-methoxy-indole-2-carboxylic acid, 4-methoxyindole-2carboxylic acid, 1h-indole-2-carboxylicacid, 4-methoxy, 1h-indole-2-carboxylic acid, 4-methoxy, 4-methoxy-indole-2-carboxylicacid, pubchem9380, acmc-20aa7f, 4-methoxy-2-indolecarboxylic acid
907248
4-methoxy-1H-indole-2-carboxylic acid
COC1=CC=CC2=C1C=C(N2)C(=O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
103260-65-7
C10H9NO3
191.186
MFCD02664458
ZZAVIQXQBBOHBB-UHFFFAOYSA-N
4-methoxyindole-2-carboxylic acid, 4-methoxy-indole-2-carboxylic acid, 4-methoxyindole-2carboxylic acid, 1h-indole-2-carboxylicacid, 4-methoxy, 1h-indole-2-carboxylic acid, 4-methoxy, 4-methoxy-indole-2-carboxylicacid, pubchem9380, acmc-20aa7f, 4-methoxy-2-indolecarboxylic acid
907248
4-methoxy-1H-indole-2-carboxylic acid
COC1=CC=CC2=C1C=C(N2)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Formula Weight 191.19
Percent Purity ≥97%
Chemical Name or Material 4-Methoxyindole-2-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'