missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxytrityl alcohol, 94%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
It is used as an intermediate for preparation of TT-0011.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an intermediate for preparation of TT-0011.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used as an intermediate for preparation of TT-0011.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Quantity | 50 g |
Solubility Information | Insoluble in water. |
Formula Weight | 290.36 |
Percent Purity | 94% |
Chemical Name or Material | 4-Methoxytrityl alcohol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'