missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-phenyl-1,3-thiazole-5-carbaldehyde, Thermo Scientific™

Catalog Number p-4510333
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
55327-23-6
C11H9NOS
203.26
MFCD04974047
QRPSQJOXOZJEHJ-UHFFFAOYSA-N
4-methyl-2-phenylthiazole-5-carbaldehyde, 4-methyl-2-phenyl-thiazole-5-carbaldehyde, 2-phenyl-4-methylthiazole-5-carbaldehyde, 5-thiazolecarboxaldehyde, 4-methyl-2-phenyl
2795491
4-methyl-2-phenyl-1,3-thiazole-5-carbaldehyde
CC1=C(SC(=N1)C1=CC=CC=C1)C=O
This item is not returnable. View return policy
55327-23-6
C11H9NOS
203.26
MFCD04974047
QRPSQJOXOZJEHJ-UHFFFAOYSA-N
4-methyl-2-phenylthiazole-5-carbaldehyde, 4-methyl-2-phenyl-thiazole-5-carbaldehyde, 2-phenyl-4-methylthiazole-5-carbaldehyde, 5-thiazolecarboxaldehyde, 4-methyl-2-phenyl
2795491
4-methyl-2-phenyl-1,3-thiazole-5-carbaldehyde
CC1=C(SC(=N1)C1=CC=CC=C1)C=O
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 203.26
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-phenyl-1, 3-thiazole-5-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'