Learn More
4-Methylimidazole, 98%, Thermo Scientific Chemicals
Catalog Number p-7027409
Click to view available options
Quantity:
10 g
50 g
250 g
822-36-6
C4H6N2
82.11
MFCD00005201
XLSZMDLNRCVEIJ-UHFFFAOYSA-N
4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol
13195
CHEBI:40035
5-methyl-1H-imidazole
CC1=CN=CN1
822-36-6
C4H6N2
82.11
MFCD00005201
XLSZMDLNRCVEIJ-UHFFFAOYSA-N
4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol
13195
CHEBI:40035
5-methyl-1H-imidazole
CC1=CN=CN1
4-Methylimidazole is used for the curing agent of epoxy resin, the main raw material of medicine cimetidine. It can also be used for synthesis of antibacterial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methylimidazole is used for the curing agent of epoxy resin, the main raw material of medicine cimetidine. It can also be used for synthesis of antibacterial agent.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY
Melting Point 49°C to 57°C
Density 1.02
Boiling Point 263°C
Flash Point 157°C (314°F)
Quantity 10 g
UN Number UN3263
Beilstein 1453
Solubility Information Soluble in water.
Formula Weight 82.11
Percent Purity 98%
Chemical Name or Material 4-Methylimidazole