missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrobenzeneboronic acid pinacol ester, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4-Nitrobenzeneboronic acid pinacol ester is a reactant involved in synthesis of aryl azides, arylation of allylic chlorides, synthesis of RNA conjugates, alkoxycarbonylation, synthesis of mTOR and PI3K inhibitors as antitumor agents, electro oxidative synthesis of biaryls.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrobenzeneboronic acid pinacol ester is a reactant involved in synthesis of aryl azides, arylation of allylic chlorides, synthesis of RNA conjugates, alkoxycarbonylation, synthesis of mTOR and PI3K inhibitors as antitumor agents, electro oxidative synthesis of biaryls.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
4-Nitrobenzeneboronic acid pinacol ester is a reactant involved in synthesis of aryl azides, arylation of allylic chlorides, synthesis of RNA conjugates, alkoxycarbonylation, synthesis of mTOR and PI3K inhibitors as antitumor agents, electro oxidative synthesis of biaryls.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Melting Point | 108°C to 109°C |
Quantity | 5 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 249.08 |
Percent Purity | 98% |
Chemical Name or Material | 4-Nitrobenzeneboronic acid pinacol ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'