missing translation for 'onlineSavingsMsg'
Learn More

4-(Pentafluorothio)phenylacetonitrile, 97%, Thermo Scientific™

Catalog Number p-7032270
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1240257-04-8
C8H6F5NS
243.195
MFCD16652419
APQCOBQUUKSALE-UHFFFAOYSA-N
4-pentafluorosulfur phenylacetonitrile, 2-4-pentafluoro-??-sulfanyl phenyl acetonitrile, 4-pentafluorothio phenylacetonitrile
66523460
2-[4-(pentafluoro-$l^{6}-sulfanyl)phenyl]acetonitrile
C1=CC(=CC=C1CC#N)S(F)(F)(F)(F)F
This item is not returnable. View return policy
1240257-04-8
C8H6F5NS
243.195
MFCD16652419
APQCOBQUUKSALE-UHFFFAOYSA-N
4-pentafluorosulfur phenylacetonitrile, 2-4-pentafluoro-??-sulfanyl phenyl acetonitrile, 4-pentafluorothio phenylacetonitrile
66523460
2-[4-(pentafluoro-$l^{6}-sulfanyl)phenyl]acetonitrile
C1=CC(=CC=C1CC#N)S(F)(F)(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3439
Formula Weight 243.19
Percent Purity 97%
Chemical Name or Material 4-(Pentafluorothio)phenylacetonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'