missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-2-(2-pyridyl)thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-7034185
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1301738-68-0
C15H10N2O2S
282.32
MFCD16251584
AFWYDGNOZTZZDG-UHFFFAOYSA-N
4-phenyl-2-2-pyridyl thiazole-5-carboxylic acid, 4-phenyl-2-pyridin-2-yl thiazole-5-carboxylic acid, 4-phenyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylic acid
73012805
4-phenyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylic acid
OC(=O)C1=C(N=C(S1)C1=CC=CC=N1)C1=CC=CC=C1
This item is not returnable. View return policy
1301738-68-0
C15H10N2O2S
282.32
MFCD16251584
AFWYDGNOZTZZDG-UHFFFAOYSA-N
4-phenyl-2-2-pyridyl thiazole-5-carboxylic acid, 4-phenyl-2-pyridin-2-yl thiazole-5-carboxylic acid, 4-phenyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylic acid
73012805
4-phenyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylic acid
OC(=O)C1=C(N=C(S1)C1=CC=CC=N1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 282.32
Percent Purity 97%
Chemical Name or Material 4-Phenyl-2-(2-pyridyl)thiazole-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'