Learn More
Biochrom 4-Position Cell Changer
4-Position Cell Changer accommodates 10 to 50mm pathlength cells. Compatible with Biochrom Ultrospec 7500/2100/3100/3300/4300 UV-Vis Spectrophotometer.
Brand: Biochrom 80210601
Additional Details : Weight : 1.00000kg
Description
- Accommodates 10 to 50 mm pathlength cells
- Compatible with Biochrom Ultrospec7500/2100/3100/3300/4300 UV-Vis Spectrophotometers
- Easy to install
Specifications
Ultrospec 2100/3100/3300/4300 UV-Vis Spectrophotometer | |
Reusable | |
10 to 50 mm |
Cell Changer | |
Biochrom Ultrospec7500/2100/3100/3300/4300 UV-Vis Spectrophotometers | |
Round |