missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrrolidinopyridine, TRC

Product Code. 30416171
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30416171 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30416171

Brand: Toronto Research Chemicals TRCP99757010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Pyrrolidinopyridine
CAS 2456-81-7
Molecular Formula C9H12N2
Quantity 10 g
Synonym 4-(1-Pyrrolidinyl)-pyridine, 4-(1-Pyrrolidino)pyridine, 4-(1-Pyrrolidinyl)pyridine, 4-(N-Pyrrolidinyl)pyridine
InChI Formula InChI=1S/C9H12N2/c1-2-8-11(7-1)9-3-5-10-6-4-9/h3-6H,1-2,7-8H2
SMILES C1CCN(C1)c2ccncc2
IUPAC Name 4-pyrrolidin-1-ylpyridine
Molecular Weight (g/mol) 148.2
Formula Weight 148.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.