Learn More
Hanna Instruments™ 4-Ring Conductivity Probes
Click to view available options
For Use With (Equipment):
EC 215
HI 8733
HI 8733, HI 9033, HI 9034, HI 87314
Unit Size:
Each
Specifications
Specifications
| Connector Types | DIN |
| For Use With (Application) | Measures electrical conductivity in liquids |
| For Use With (Equipment) | HI 8733, HI 9033, HI 9034, HI 87314 |
| Type | Multi-range Portable EC Meter |
| Length (Metric) | 140 mm |
| Length (Metric) Cable | 1 m |
| Temperature Sensor | Yes |