missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzonitrile, 98+%, Thermo Scientific Chemicals

Catalog Number p-7021658
18.40 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4210-32-6
C11H13N
159.232
MFCD00075840
IIZURLNRIMKEDL-UHFFFAOYSA-N
4-tert-butyl benzonitrile, 4-t-butylbenzonitrile, benzonitrile, 4-1,1-dimethylethyl, 4-tert-butylcyanobenzene, 4-tert-butyl-benzonitrile, p-t-butylbenzonitrile, acmc-209jnk, p-tert-butylbenzonitrile, benzonitrile, p-tert-butyl, ksc219k6d
77883
4-tert-butylbenzonitrile
CC(C)(C)C1=CC=C(C=C1)C#N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4210-32-6
C11H13N
159.232
MFCD00075840
IIZURLNRIMKEDL-UHFFFAOYSA-N
4-tert-butyl benzonitrile, 4-t-butylbenzonitrile, benzonitrile, 4-1,1-dimethylethyl, 4-tert-butylcyanobenzene, 4-tert-butyl-benzonitrile, p-t-butylbenzonitrile, acmc-209jnk, p-tert-butylbenzonitrile, benzonitrile, p-tert-butyl, ksc219k6d
77883
4-tert-butylbenzonitrile
CC(C)(C)C1=CC=C(C=C1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.94
Boiling Point 258°C
Flash Point >110°C (230°F)
Odor Characteristic
Refractive Index 1.517
Quantity 1 g
Beilstein 2080049
Formula Weight 159.23
Percent Purity ≥98%
Chemical Name or Material 4-tert-Butylbenzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'