Learn More
Alfa Aesar™ 4-(trans-3-Methoxy-3-oxo-1-propen-1-yl)benzeneboronic acid, 97%
Brand: Alfa Aesar™ H53292.MD
Additional Details : CAS : 380430-58-0 Weight : 0.00025kg
Quantity | 250mg |
---|
Description
Applications4-(trans-3-Methoxy-3-oxo-1-propen-1-yl)benzeneboronic acid used as reagents for suzuki-miyaura cross-coupling reacion.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerate.
Chemical Identifiers
380430-58-0 | |
206.004 | |
BIKAXBPCHWDATP-QPJJXVBHSA-N | |
5708389 | |
B(C1=CC=C(C=C1)C=CC(=O)OC)(O)O |
C10H11BO4 | |
MFCD02179476 | |
4-e-3-methoxy-3-oxo-1-propen-1-yl phenyl boronic acid, 4-3-methoxy-3-oxoprop-1-en-1-yl phenyl boronic acid, 4-e-3-methoxy-3-oxo-1-propen-1-yl phenylboronic acid, 4-trans-3-methoxy-3-oxo-1-propen-1-yl benzeneboronic acid, 4-3-methoxy-3-oxoprop-1-enyl phenylboronic acid, 4-1e-3-methoxy-3-oxoprop-1-en-1-yl phenylboronic acid, 4-3-methoxy-3-oxoprop-1-en-1-yl phenylboronic acid, 4-e-3-methoxy-3-oxoprop-1-enyl phenyl boronic acid, 4-1e-3-methoxy-3-oxoprop-1-en-1-yl phenyl boronic acid, 2-propenoic acid, 3-4-boronophenyl-, 1-methyl ester, 2e | |
[4-[(E)-3-methoxy-3-oxoprop-1-enyl]phenyl]boronic acid |
Specifications
4-(trans-3-Methoxy-3-oxo-1-propen-1-yl)benzeneboronic acid | |
250mg | |
C10H11BO4 | |
4-e-3-methoxy-3-oxo-1-propen-1-yl phenyl boronic acid, 4-3-methoxy-3-oxoprop-1-en-1-yl phenyl boronic acid, 4-e-3-methoxy-3-oxo-1-propen-1-yl phenylboronic acid, 4-trans-3-methoxy-3-oxo-1-propen-1-yl benzeneboronic acid, 4-3-methoxy-3-oxoprop-1-enyl phenylboronic acid, 4-1e-3-methoxy-3-oxoprop-1-en-1-yl phenylboronic acid, 4-3-methoxy-3-oxoprop-1-en-1-yl phenylboronic acid, 4-e-3-methoxy-3-oxoprop-1-enyl phenyl boronic acid, 4-1e-3-methoxy-3-oxoprop-1-en-1-yl phenyl boronic acid, 2-propenoic acid, 3-4-boronophenyl-, 1-methyl ester, 2e | |
BIKAXBPCHWDATP-QPJJXVBHSA-N | |
[4-[(E)-3-methoxy-3-oxoprop-1-enyl]phenyl]boronic acid | |
5708389 | |
97% |
234°C to 240°C | |
380430-58-0 | |
MFCD02179476 | |
Slightly soluble in water. | |
B(C1=CC=C(C=C1)C=CC(=O)OC)(O)O | |
206.004 | |
206 |