Learn More
BRAND™ BRANDplates™ Microplate Lids
Shop All Brand Products
Click to view available options
Plate Compatibility:
1536-well Standard Plate
384-well Plate
96-well Plate
96-well Standard Plate
No. per Pack:
50
100
Condensation Rings:
No
Yes
Height (Metric):
4.5 mm
5.5 mm
8 mm
Unit Size:
Pack of 100
Pack of 50
Description
- Made from crystal-clear polystyrene
- Cut-corner sets orientation of lid
- Stackable
Lids with condensation rings available for 96-well standard plates
Specifications
Specifications
| Color | Transparent |
| Material | Polystyrene |
| Sterility | Non-sterile |
| No. of Wells | 384 |
| Height (Metric) | 4.5 mm |
| Condensation Rings | No |
| No. per Pack | 50 |
| Evaporation Barrier | No |
| Product Type | Microplate Cover |
| Adhesive | None |
| Show More |