missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Nitrophenyl)-2-furoyl chloride, Thermo Scientific™

Product Code. 11375409
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11375409

Brand: Thermo Scientific Alfa Aesar H50406.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61941-88-6
Molecular Formula C11H6ClNO4
Molecular Weight (g/mol) 251.622
MDL Number MFCD02258036
InChI Key RRWHGDGMPQFABA-UHFFFAOYSA-N
Synonym 5-2-nitrophenyl furan-2-carbonyl chloride, 5-2-nitrophenyl-2-furoyl chloride, 5-2-nitro-phenyl-furan-2-carbonyl chloride, acmc-20am9n, 5-2-nitrophenyl furan-2-carboxylic acid chloride
PubChem CID 2771973
IUPAC Name 5-(2-nitrophenyl)furan-2-carbonyl chloride
SMILES C1=CC=C(C(=C1)C2=CC=C(O2)C(=O)Cl)[N+](=O)[O-]

Specifications

Quantity 1 g
UN Number UN3261
Formula Weight 251.63
Chemical Name or Material 5-(2-Nitrophenyl)-2-furoyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.