missing translation for 'onlineSavingsMsg'
Learn More

5,6-Diphenyl-1,2,4-triazin-3-amine, 97%, Thermo Scientific™

Catalog Number p-2702451
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
4511-99-3
C15H12N4
248.289
MFCD00047461
NZRHOWNFGASHMN-UHFFFAOYSA-N
78268
5,6-diphenyl-1,2,4-triazin-3-amine
C1=CC=C(C=C1)C2=C(N=NC(=N2)N)C3=CC=CC=C3
This item is not returnable. View return policy
4511-99-3
C15H12N4
248.289
MFCD00047461
NZRHOWNFGASHMN-UHFFFAOYSA-N
78268
5,6-diphenyl-1,2,4-triazin-3-amine
C1=CC=C(C=C1)C2=C(N=NC(=N2)N)C3=CC=CC=C3
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-amino-5,6-diphenyl-1,2,4-triazine, diphenyl-1,2,4-triazin-3-amine, as-triazine, 3-amino-3,6-diphenyl, 5,6-diphenyl-1,2,4-triazin-3-ylamine, 3-amino-5,6-diphenyl-as-triazine, as-triazine, 3-amino-5,6-diphenyl, 1,2,4-triazin-3-amine,5,6-diphenyl, 1,2,4-triazin-3-amine, 5,6-diphenyl, 5,6-diphenyl-1,2,4-triazin-3 2h-imine, 5,6-diphenyl-1,2,4-triazine-3-ylamine
Formula Weight 248.29
Percent Purity 97%
Chemical Name or Material 5,6-Diphenyl-1,2,4-triazin-3-amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'