missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 5,7-Dichlorokynurenic acid sodium salt
GREENER_CHOICE

Product Code. 18024082
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18024082

Brand: Tocris Bioscience™ 3698/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium salt of 5,7-Dichlorokynurenic acid (Cat. No. 0286). Potent NMDA antagonist, acts at glycine site

5,7-Dichlorokynurenic acid sodium salt is a sodium salt of 5,7-Dichlorokynurenic acid (Cat. No. 0286), a potent antagonist at the glycine site of the NMDA receptor (Ki = 79 nM vs. [3H]-glycine).
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type 5,7-Dichlorokynurenic acid sodium salt
Purity >98%
Molecular Weight (g/mol) 280.04
Inhibitors 5,7-Dichlorokynurenic acid sodium salt

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.