missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-methyl-1,3-benzoxazole, 97%, Thermo Scientific Chemicals

Catalog Number p-4228325
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
72745-76-7
C8H8N2O
148.17
MFCD00628970
FECYTFWPNCBIHC-UHFFFAOYSA-N
2-methylbenzo d oxazol-5-amine, 2-methyl-5-aminobenzoxazole, 5-amino-2-methylbenzoxazole, 5-benzoxazolamine,2-methyl, 2-methylbenzoxazole-5-ylamine, 2-methyl-benzoxazol-5-ylamine, 5-benzoxazolamine, 2-methyl, 2-methyl-benzooxazol-5-ylamine, 2-methylbenzoxazol-5-yl amine, 5-amino-2-methyl-1,3-benzoxazole
737920
2-methyl-1,3-benzoxazol-5-amine
CC1=NC2=CC(N)=CC=C2O1
This item is not returnable. View return policy
72745-76-7
C8H8N2O
148.17
MFCD00628970
FECYTFWPNCBIHC-UHFFFAOYSA-N
2-methylbenzo d oxazol-5-amine, 2-methyl-5-aminobenzoxazole, 5-amino-2-methylbenzoxazole, 5-benzoxazolamine,2-methyl, 2-methylbenzoxazole-5-ylamine, 2-methyl-benzoxazol-5-ylamine, 5-benzoxazolamine, 2-methyl, 2-methyl-benzooxazol-5-ylamine, 2-methylbenzoxazol-5-yl amine, 5-amino-2-methyl-1,3-benzoxazole
737920
2-methyl-1,3-benzoxazol-5-amine
CC1=NC2=CC(N)=CC=C2O1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 72°C to 74.6°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 148.17
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-methyl-1, 3-benzoxazole
Show More Show Less
compliance-icons
Product Identifier
  • 5-Amino-2-methyl-1,3-benzoxazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'