missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-phenylisoxazole, 97%, Thermo Scientific™

Catalog Number p-4228334
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4369-55-5
C9H8N2O
160.18
MFCD00464114
HLOHVVZZMMMDMM-UHFFFAOYSA-N
3-phenylisoxazol-5-amine, 5-amino-3-phenylisoxazole, 5-isoxazolamine, 3-phenyl, 3-phenyl-5-isoxazolamine, 3-phenylisoxazole-5-ylamine, isoxazole, 5-amino-3-phenyl, phenylisoxazolamine, pubchem15538, 3-phenylisoxazole-5-amine, acmc-209jv8
261201
3-phenyl-1,2-oxazol-5-amine
NC1=CC(=NO1)C1=CC=CC=C1
This item is not returnable. View return policy
4369-55-5
C9H8N2O
160.18
MFCD00464114
HLOHVVZZMMMDMM-UHFFFAOYSA-N
3-phenylisoxazol-5-amine, 5-amino-3-phenylisoxazole, 5-isoxazolamine, 3-phenyl, 3-phenyl-5-isoxazolamine, 3-phenylisoxazole-5-ylamine, isoxazole, 5-amino-3-phenyl, phenylisoxazolamine, pubchem15538, 3-phenylisoxazole-5-amine, acmc-209jv8
261201
3-phenyl-1,2-oxazol-5-amine
NC1=CC(=NO1)C1=CC=CC=C1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 105.9°C to 109.8°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 160.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-3-phenylisoxazole
Show More Show Less
compliance-icons
Product Identifier
  • 5-Amino-3-phenylisoxazole, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'