missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-phenyl-7-azaindole, 97%, Thermo Scientific Chemicals

Catalog Number 15407487
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
953414-75-0
C13H9BrN2
273.13
MFCD11109822
QLKZJMXSXCDCCF-UHFFFAOYSA-N
5-bromo-2-phenyl-1h-pyrrolo 2,3-b pyridine, 5-bromo-2-phenyl-7-azaindole, 1h-pyrrolo 2,3-b pyridine,5-bromo-2-phenyl, 2-phenyl-5-bromo-1h-pyrrolo 2,3-b pyridine
23643607
5-bromo-2-phenyl-1H-pyrrolo[2,3-b]pyridine
BrC1=CN=C2NC(=CC2=C1)C1=CC=CC=C1
This item is not returnable. View return policy
953414-75-0
C13H9BrN2
273.13
MFCD11109822
QLKZJMXSXCDCCF-UHFFFAOYSA-N
5-bromo-2-phenyl-1h-pyrrolo 2,3-b pyridine, 5-bromo-2-phenyl-7-azaindole, 1h-pyrrolo 2,3-b pyridine,5-bromo-2-phenyl, 2-phenyl-5-bromo-1h-pyrrolo 2,3-b pyridine
23643607
5-bromo-2-phenyl-1H-pyrrolo[2,3-b]pyridine
BrC1=CN=C2NC(=CC2=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 287°C to 289°C
Color Brown
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 273.12
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-phenyl-7-azaindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'