missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2,4-dimethylbenzene-1-thiol, TRC

Product Code. 30350958
Change view
Click to view available options
Quantity:
250 mg
Product Code. Quantity
30350958 250 mg
1 options
This item is not returnable. View return policy

Product Code. 30350958

Brand: Toronto Research Chemicals TRCC371545250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chloro-2,4-dimethylbenzene-1-thiol
CAS 1823050-95-8
Molecular Formula C8H9ClS
Quantity 250 mg
InChI Formula InChI=1S/C8H9ClS/c1-5-3-6(2)8(10)4-7(5)9/h3-4,10H,1-2H3
SMILES Cc1cc(C)c(Cl)cc1S
IUPAC Name 5-chloro-2,4-dimethylbenzenethiol
Molecular Weight (g/mol) 172.68
Formula Weight 172.0113
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.