missing translation for 'onlineSavingsMsg'
Learn More

5-Cyano-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15407806
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
863868-29-5
C13H15BFNO2
247.08
MFCD07437899
DCBKFUVWBRFDDD-UHFFFAOYSA-N
4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 5-cyano-2-fluorophenylboronic acid, pinacol ester, 5-cyano-2-fluorobenzeneboronic acid pinacol ester, 5-cyano-2-fluorophenylboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-4-fluorobenzonitrile, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 4-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, benzonitrile, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
53426612
4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
CC1(C)OB(OC1(C)C)C1=C(F)C=CC(=C1)C#N
This item is not returnable. View return policy
863868-29-5
C13H15BFNO2
247.08
MFCD07437899
DCBKFUVWBRFDDD-UHFFFAOYSA-N
4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 5-cyano-2-fluorophenylboronic acid, pinacol ester, 5-cyano-2-fluorobenzeneboronic acid pinacol ester, 5-cyano-2-fluorophenylboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-4-fluorobenzonitrile, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 4-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, benzonitrile, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
53426612
4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
CC1(C)OB(OC1(C)C)C1=C(F)C=CC(=C1)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 247.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 5-Cyano-2-fluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'