missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 5-Ethynyluridine

Description
5-Ethynyluridine is a cell-permeable uridine analog. 5-Ethynyluridine can be used in vitro and in vivo to label newly synthesized RNA. 5-Ethynyluridine can be used to study changes in transcription.
Specifications
Specifications
| Chemical Name or Material | 1-((2 R,3 R,4 S,5 R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-ethynylpyrimidine-2,4(1 H,3 H)-dione |
| CAS | 69075-42-9 |
| Quantity | 10 mg |
| Target | DNA, RNA and Protein Synthesis Compounds |
| Molecular Formula | C11H12N2O6 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?