Learn More
5-Fluorosalicylic acid, 97%
Product Code. p-7025990
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 11324659
Brand: Thermo Scientific Alfa Aesar B21751.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
CAS: 345-16-4 | C7H5FO3 | 156.112 g/mol
5-Fluorosalicylic acid was used as internal standard in the quantification of salicylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Fluorosalicylic acid was used as internal standard in the quantification of salicylic acid.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 345-16-4
Molecular Formula C7H5FO3
Molecular Weight (g/mol) 156.112
MDL Number MFCD00002456
InChI Key JWPRICQKUNODPZ-UHFFFAOYSA-N
Synonym 5-fluorosalicylic acid, 2-hydroxy-5-fluorobenzoic acid, benzoic acid, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxybenzoicacid, 5-fluoro-2-hydroxy-benzoic acid, 5fluorosalicylic acid, pubchem1382, 5-fluoro-salicylic acid, 2-carboxy-4-fluorophenol
PubChem CID 67658
IUPAC Name 5-fluoro-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1F)C(=O)O)O
Specifications
Melting Point 178°C to 182°C
Quantity 250 mg
Beilstein 2209120
Solubility Information Slightly soluble in water.
Formula Weight 156.11
Percent Purity 97%
Chemical Name or Material 5-Fluorosalicylic acid