missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorouracil, 99%, Thermo Scientific Chemicals

Catalog Number p-7021215
Click to view available options
Quantity:
5 g
25 g
100 g
51-21-8
C4H3FN2O2
130.08
MFCD00006018
GHASVSINZRGABV-UHFFFAOYSA-N
5-fluorouracil, fluorouracil, 5-fu, fluoroplex, adrucil, efudex, carac, fluracil, fluoroblastin, kecimeton
3385
CHEBI:46345
5-fluoro-1H-pyrimidine-2,4-dione
FC1=CNC(=O)NC1=O
51-21-8
C4H3FN2O2
130.08
MFCD00006018
GHASVSINZRGABV-UHFFFAOYSA-N
5-fluorouracil, fluorouracil, 5-fu, fluoroplex, adrucil, efudex, carac, fluracil, fluoroblastin, kecimeton
3385
CHEBI:46345
5-fluoro-1H-pyrimidine-2,4-dione
FC1=CNC(=O)NC1=O

5-Fluorouracil is used as an antitumor agent in the treatment of anal, breast, colorectal, oesophageal, stomach, pancreatic and skin cancers. It finds application as a suicide inhibitor due to its irreversible inhibition of thymidylate synthase. It is also used in the treatment of actinic keratoses and bowen's disease. Further, it serves as a potent antineoplastic agent in clinical use. In addition to this, it acts as a DNA synthesis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethylformamide, dimethyl sulfoxide, methanol, hot water, ammonium hydroxide, sodium hydroxide, ethanol and water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Melting Point ∽282°C (decomposition)
Assay Percent Range 99%
Quantity 5 g
UN Number UN2811
Beilstein 127172
Sensitivity Light sensitive
Merck Index 14,4181
Solubility Information Soluble in dimethylformamide,dimethyl sulfoxide,methanol,hot water,ammonium hydroxide,sodium hydroxide,ethanol and water.
Formula Weight 130.08
Percent Purity 99%
Chemical Name or Material 5-Fluorouracil
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Fluorouracil
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Fluorouracil

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'