missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hexynoic Acid, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 5-Hexynoic Acid |
| CAS | 53293-00-8 |
| Molecular Formula | C6H8O2 |
| Quantity | 100 mg |
| Synonym | 4-Ethynylbutyric Acid, 5-Hexyn-1-oic Acid |
| InChI Formula | InChI=1S/C6H8O2/c1-2-3-4-5-6(7)8/h1H,3-5H2,(H,7,8) |
| SMILES | OC(=O)CCCC#C |
| IUPAC Name | hex-5-ynoic acid |
| Molecular Weight (g/mol) | 112.13 |
| Formula Weight | 112.0524 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction