missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 5-InoAz
GREENER_CHOICE

Product Code. p-200079397
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18784374

Brand: Tocris Bioscience™ 7973/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Metabolic probe for labeling mycobacterium cell envelope

5-InoAz is a metabolic probe for labeling the mycobacterium cell envelope. Specifically designed as an inositol analog to incorportate into cell envelope glycolipids (phosphatidylinositol mannoside (PIM), PIM-anchored lipomannan (LM) and lipoarabinomannan (LAM)). In combination with bioorthogonal chemistry, it provides a valuable tool for investigating PIM, LM, and LAM biosynthesis, transport, and dynamics in diverse mycobacterial organisms.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Azido-5-deoxy-myo-inositol
CAS 1218932-97-8
Quantity 50 mg
Target Glycobiology Probes
Molecular Formula C6H11N3O5
Purity 0.9
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.