missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-(trifluoromethyl)-3-furoic acid, 97%, Thermo Scientific™

Catalog Number p-4509798
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
17515-74-1
C7H5F3O3
194.11
MFCD00275563
NURNFMNHTHXTDS-UHFFFAOYSA-N
2782183
5-methyl-2-(trifluoromethyl)furan-3-carboxylic acid
CC1=CC(C(O)=O)=C(O1)C(F)(F)F
This item is not returnable. View return policy
17515-74-1
C7H5F3O3
194.11
MFCD00275563
NURNFMNHTHXTDS-UHFFFAOYSA-N
2782183
5-methyl-2-(trifluoromethyl)furan-3-carboxylic acid
CC1=CC(C(O)=O)=C(O1)C(F)(F)F
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Synonym 5-methyl-2-trifluoromethyl furan-3-carboxylic acid, 5-methyl-2-trifluoromethyl-3-furoic acid, 5-methyl-2-trifluoromethyl-furan-3-carboxylic acid, 2-trifluoromethyl-5-methyl-3-furancarboxylic acid, 3-furancarboxylic acid, 5-methyl-2-trifluoromethyl, 2-trifluoromethyl-5-methyl-3-furoic acid, 2-trifluoromethyl-5-methylfuran-3-carboxylic acid, 5-methyl-2-trifluoromethyl-3-furancarboxylic acid, 3-furancarboxylic acid,5-methyl-2-trifluoromethyl
Formula Weight 194.11
Percent Purity 97%
Chemical Name or Material 5-Methyl-2-(trifluoromethyl)-3-furoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'