missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-5-phenylhydantoin, 99%, Thermo Scientific Chemicals

Catalog Number p-3758320
26.46 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6843-49-8
C10H10N2O2
190.2
MFCD00005265
JNGWGQUYLVSFND-UHFFFAOYSA-N
93043
5-methyl-5-phenylimidazolidine-2,4-dione
CC1(C(=O)NC(=O)N1)C2=CC=CC=C2
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6843-49-8
C10H10N2O2
190.2
MFCD00005265
JNGWGQUYLVSFND-UHFFFAOYSA-N
93043
5-methyl-5-phenylimidazolidine-2,4-dione
CC1(C(=O)NC(=O)N1)C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 199°C to 201°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 24, I, 347
Synonym 5-methyl-5-phenylhydantoin, hydantoin, 5-methyl-5-phenyl, 2,4-imidazolidinedione,5-methyl-5-phenyl, 2,4-imidazolidinedione, 5-methyl-5-phenyl, mpid standard, acmc-20m4pt, 2,4-imidazolidinedione, 5-methyl-5-phenyl-, r, 2, 5-methyl-5-phenyl, +/--5-methyl-5-phenylhydantoin
Formula Weight 190.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-5-phenylhydantoin
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'