missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylsimvastatin, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
5-Methylsimvastatin
Specifications
Specifications
| Chemical Name or Material | 2-Methyl Simvastatin (mixture of diasteroisomers) |
| CAS | 774611-54-0 |
| Analyte or Component Names | 2-Methyl Simvastatin (mixture of diasteroisomers) |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C26 H40 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Butanoic acid, 2,2-dimethyl-, (1S,3R,7S,8S,8aR)-1,2,3,7,8,8a-hexahydro-3,7-dimethyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-5-methyl-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl ester, (1S,3R,7S,8S,8aR)-1,2,3,7,8,8a-Hexahydro-3,7-dimethyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-5-methyl-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl 2,2-dimethylbutanoate, 5-Methylsimvastatin, (1S,3R,7S,8S,8aR)-8-[2-[(2R,4R,5RS)-4-Hydroxy-5-methyl-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2,2-Dimethylbutanoate |
| InChI Formula | InChI=1S/C26H40O5/c1-7-26(5,6)25(29)31-22-13-15(2)12-18-9-8-16(3)20(23(18)22)11-10-19-14-21(27)17(4)24(28)30-19/h8-9,12,15-17,19-23,27H,7,10-11,13-14H2,1-6H3/t15-,16-,17?,19+,20-,21+,22-,23-/m0/s1 |
| SMILES | CCC(C)(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)C(C)C(=O)O3)[C@@H]12 |
| IUPAC Name | [(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-hydroxy-5-methyl-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] 2,2-dimethylbutanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction