Learn More
5-O-tert-Butyldiphenylsilyl-2,3-O-isopropylidene-alpha,beta-D-ribofuranose, Thermo Scientific Chemicals
Catalog Number p-7046711
Click to view available options
Quantity:
1 g
5 g
141607-35-4
C24H32O5Si
428.6
MFCD01075719
OOVHEXLXHQNNLL-RQLHVLPXSA-N
133612129
(3aS,6S,6aS)-6-[[tert-butyl(diphenyl)silyl]oxymethyl]-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-ol
CC1(OC2C(OC(C2O1)O)CO[Si](C3=CC=CC=C3)(C4=CC=CC=C4)C(C)(C)C)C
141607-35-4
C24H32O5Si
428.6
MFCD01075719
OOVHEXLXHQNNLL-RQLHVLPXSA-N
133612129
(3aS,6S,6aS)-6-[[tert-butyl(diphenyl)silyl]oxymethyl]-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-ol
CC1(OC2C(OC(C2O1)O)CO[Si](C3=CC=CC=C3)(C4=CC=CC=C4)C(C)(C)C)C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Optical Rotation −28° (c=1.5 in Chloroform)
Formula Weight 428.6
Chemical Name or Material 5-O-tert-Butyldiphenylsilyl-2,3-O-isopropylidene-alpha,beta-D-ribofuranose