missing translation for 'onlineSavingsMsg'
Learn More

5-(o-Tolyl)-1,3,4-oxadiazole-2-thiol, 96%, Thermo Scientific™

Catalog Number 15406637
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2503-66-4
C9H8N2OS
192.236
MFCD00992514
HNFIJWNVPWXSOL-UHFFFAOYSA-N
5-2-methylphenyl-1,3,4-oxadiazole-2-thiol, 5-o-tolyl-3h-1,3,4 oxadiazole-2-thione, 5-o-tolyl-1,3,4-oxadiazole-2-thiol, 5-2-methylphenyl-3h-1,3,4-oxadiazole-2-thione, 1,3,4-oxadiazole-2 3h-thione,5-2-methylphenyl, 5-o-tolyl-3h-1,3,4-oxadiazole-2-thione, 5-2-methylphenyl-1,3,4-oxadiazoline-2-thione, 5-2-methylphenyl-1,3,4-oxadiazol-2 3h-thione
774385
5-(2-methylphenyl)-3H-1,3,4-oxadiazole-2-thione
CC1=CC=CC=C1C2=NNC(=S)O2
This item is not returnable. View return policy
2503-66-4
C9H8N2OS
192.236
MFCD00992514
HNFIJWNVPWXSOL-UHFFFAOYSA-N
5-2-methylphenyl-1,3,4-oxadiazole-2-thiol, 5-o-tolyl-3h-1,3,4 oxadiazole-2-thione, 5-o-tolyl-1,3,4-oxadiazole-2-thiol, 5-2-methylphenyl-3h-1,3,4-oxadiazole-2-thione, 1,3,4-oxadiazole-2 3h-thione,5-2-methylphenyl, 5-o-tolyl-3h-1,3,4-oxadiazole-2-thione, 5-2-methylphenyl-1,3,4-oxadiazoline-2-thione, 5-2-methylphenyl-1,3,4-oxadiazol-2 3h-thione
774385
5-(2-methylphenyl)-3H-1,3,4-oxadiazole-2-thione
CC1=CC=CC=C1C2=NNC(=S)O2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 181°C to 182°C
Quantity 5 g
Formula Weight 192.24
Percent Purity 96%
Chemical Name or Material 5-(o-Tolyl)-1,3,4-oxadiazole-2-thiol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'