missing translation for 'onlineSavingsMsg'
Learn More

TdB Labs AB 500 kDa CM-dextran Carboxymethylated for Cation Exchange and Gel Electrophoresis

Product Code. 30216434
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
Product Code. Quantity unitSize
30216434 100 g 100g
30216258 10 g 10g
2 options
This item is not returnable. View return policy

Product Code. 30216434

Brand: TdB Labs AB CMD500_100g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CM-dextran 500 kDa is a high molecular weight, carboxymethylated polysaccharide used in viscoelastic systems, hydrogel matrices, and as a cation exchanger in electrophoresis. Its size and charge support structural and separation-based applications.

  • Product Overview: CM-dextrans are carboxymethylated derivatives of dextran, designed for use in a variety of biological and chemical assays. Available in molecular weights ranging from 5 kDa to 2000 kDa, CM-dextrans are widely used for permeability studies, cell surface modification, and as stabilizers in drug delivery systems.
  • Features:
    • Carboxymethylation enhances solubility and biocompatibility, making CM-dextrans ideal for use in both in vitro and in vivo studies.
    • Available in a broad range of molecular weights from 5 kDa to 2000 kDa, providing flexibility for different experimental needs.
    • Water-soluble, facilitating incorporation into aqueous biological systems for assays and molecular tracking.
    • Stable and non-toxic, ensuring minimal interference with biological systems during experiments.
  • Benefits:
    • Versatile Molecular Weights: Available in 5, 10, 20, 40, 70, 150, 500, and 2000 kDa, CM-dextrans allow for precise control over molecular size and diffusion properties in permeability, drug delivery, and other applications.
    • Enhanced Solubility: The carboxymethylation process improves solubility, ensuring smooth integration into cell culture systems and biological assays.
    • Biocompatible and Safe: CM-dextrans are safe for live-cell applications and in vivo studies, offering low toxicity and minimal disruption to biological systems.
    • Stability and Reproducibility: CM-dextrans provide consistent results across experiments, offering long-term stability for storage and reliable performance in assays.
  • Applications:
    • Used in permeability assays to evaluate the transport of molecules across cell monolayers and biological barriers.
    • Commonly used in drug delivery research to stabilize therapeutic agents and improve their delivery to target tissues.
    • Effective in cell surface modification for creating functionalized surfaces in biomedical applications.
    • Applied in vascular research to study macromolecular transport and capillary permeability in disease models.
    • Used in hydrogel formation to create crosslinked networks for tissue engineering and regenerative medicine.
  • Technical Specifications:
    • Molecular Weights: Available in 5 kDa, 10 kDa, 20 kDa, 40 kDa, 70 kDa, 150 kDa, 500 kDa, and 2000 kDa to fit different experimental needs.
    • Solubility: Fully soluble in water for easy incorporation into biological systems.
    • Carboxymethylation: Improves solubility and biocompatibility, allowing for use in a wide range of biological applications.
  • Storage and Stability:
    • Stable for extended periods when stored dry in sealed containers at ambient temperature.
    • Protect from moisture and light to maintain optimal stability and effectiveness over time.
  • Packaging and Delivery:
    • CM-dextrans are supplied as powders, ensuring long shelf life and easy handling in laboratory settings.
    • Available in multiple packaging sizes, ranging from small quantities for initial research to bulk packaging for larger studies.

Specifications

Product Type Dextran
Form Powder
Quantity 100 g
Molecular Weight (g/mol) 500 kDa
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.