missing translation for 'onlineSavingsMsg'
Learn More

3,5,3',5'-Tetraiodo Thyroacetic Acid, TRC

Product Code. 30414159
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 30414159

Brand: Toronto Research Chemicals TRCT296125500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5,3',5'-Tetraiodo Thyroacetic Acid
CAS 67-30-1
Purity Grade Notes HPLC
Molecular Formula C14 H8 I4 O4
Quantity 500 mg
Synonym Benzeneacetic acid','4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodo-, Acetic acid','[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]- (8CI), Thyroacetic acid','3,3',5,5'-tetraiodo- (6CI), 3,3',5,5'-Tetraiodothyroacetic acid, 3,5,3',5'-Tetraiodothyroacetic acid, Tetrac, Tetraiodothyroacetic acid, Liothyronine Sodium Imp. D (EP)
InChI Formula InChI=1S/C14H8I4O4/c15-8-4-7(5-9(16)13(8)21)22-14-10(17)1-6(2-11(14)18)3-12(19)20/h1-2,4-5,21H,3H2,(H,19,20)
SMILES OC(=O)Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1
IUPAC Name 2-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]acetic acid
Molecular Weight (g/mol) 747.83
Formula Weight 747.6601
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.