Learn More
Benzophenone-d10, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Benzophenone-d10 |
| CAS | 22583-75-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C13 D10 O |
| Quantity | 500 mg |
| Synonym | Methanone, di(phenyl-2,3,4,5,6-d5)- (ACI), Benzophenone-d10 (7CI, 8CI), Di(phenyl-2,3,4,5,6-d5)methanone (ACI), Methanone, di(phenyl-d5)- (9CI), Bis(pentadeuterophenyl)methanone, Decadeuteriobenzophenone, Perdeuteriobenzophenone, Perdeuterobenzophenone, Benzophenone-D10 |
| InChI Formula | InChI=1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D |
| SMILES | [2H]c1c([2H])c([2H])c(C(=O)c2c([2H])c([2H])c([2H])c([2H])c2[2H])c([2H])c1[2H] |
| IUPAC Name | bis(2,3,4,5,6-pentadeuteriophenyl)methanone |
| Molecular Weight (g/mol) | 192.2795 |
| Show More |