Learn More
Chem-Lab 500ML Arsenicum standard solution (Spectro ECON)(1.32 g As2O3 / l 2% HCl), for AAS
Brand: Chem-Lab CL01.0137.0500
500ML Arsenicum standard solution (Spectro ECON)(1.32 g As2O3 / l 2% HCl), for AAS
Safety and Handling
- Arsenic standard solution 1000 µg/ml
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 1A
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H350-May cause cancer.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Hydrochloric acid, Arsenic(III) oxide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.