Learn More
Chromate Standard: CrO4(-2) @ 1000 μg/mL in H2O, VHG™
Product Code. 30002931
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 30002931
Brand: VHG Labs VHGICRO500
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Chromate Standard: CrO4(-2) @ 1000 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Spécification
Chemical Name or Material Chromate
CAS 11104-59-9
Methods Ion Chromatography/Wet Chemistry
Analyte or Component Names Chromate
Shipping Condition Room Temperature
Molecular Formula (Cr O4)2-
Synonym CrO42−
SMILES [O-][Cr](=O)(=O)[O-]
IUPAC Name Chromate
Solvent or Matrix Water
Molecular Weight (g/mol) 115.99
Quantity 500 mL
Formula Weight 115.99 g/mol