Learn More
Reagecon 500ML ICME (4) Cation Standard Ca@1500 ppm, Mg@500 ppm, Na@500 ppm, K@250ppm in Water
Brand:  Reagecon ICCMX05
500ML ICME (4) Cation Standard Ca@1500 ppm, Mg@500 ppm, Na@500 ppm, K@250ppm in Water
Product Code. 16450834
  • ONLINE EXCLUSIVE PRICE
    512.00€
    413.08€ / 500mL
    Save 98.92€
    19% Off